Learn More
Honeywell™ TRICONYL Microdots Gloves
Click to view available options
Size:
8
10
Description
- Polyamide knitted glove
- Small black PVC dots on the palm
- The knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
- The glove offers maximum comfort and added sturdiness
- Non pilling, the polyamide fabric is suitable for the agrifood industry as well as for precision work
- The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The PVC dots ensure greater abrasion resistance and a better grip
Specifications
Specifications
Certifications/Compliance | EN 420-03 EEC Basic standards |
Description | 13 gauge |
Coating Material | Polyamide |
Size | 8 |
Length (Metric) | 26 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Honeywell™ TRICONYL Microdots Gloves >
Spot an opportunity for improvement?Share a Content Correction