missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetamidine, tech. 85%

Product Code. 11338795
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11338795

Brand: Thermo Scientific Alfa Aesar L12422.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 354-37-0 | C2H3F3N2 | 112.06 g/mol

Trifluoroacetamidine is used as probe for determination of membrane potential and extra/intracellular volume of erythrocytes by fluorine-19 NMR studies. It is a reagent used as protected ammonia equivalent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trifluoroacetamidine is used as probe for determination of membrane potential and extra/intracellular volume of erythrocytes by fluorine-19 NMR studies. It is a reagent used as protected ammonia equivalent.

Solubility
Fully miscible in water.

Notes
Store at -20°C. Incompatible with oxidizing agents. Moisture Sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 354-37-0
Molecular Formula C2H3F3N2
Molecular Weight (g/mol) 112.06
MDL Number MFCD00041545
InChI Key NITMACBPVVUGOJ-UHFFFAOYSA-N
Synonym trifluoroacetamidine, trifluoroethanimidamide, 2,2,2-trifluoroacetamidine, ethanimidamide, 2,2,2-trifluoro, sntljqhjptadtaetp@, trifluoroacetoamidine, trifluoroacetamideine, trifluoro-acetamidine, acmc-20aje4, 2,2,2-trifluoroacetoamidine
PubChem CID 2776882
IUPAC Name 2,2,2-trifluoroethanimidamide
SMILES NC(=N)C(F)(F)F

Specifications

Density 1.45
Boiling Point 35°C to 37°C (11 mmHg)
Refractive Index 1.382
Quantity 25 g
UN Number UN3267
Beilstein 605978
Sensitivity Moisture sensitive
Solubility Information Fully miscible in water.
Formula Weight 112.05
Percent Purity 85%
Grade Technical
Chemical Name or Material Trifluoroacetamidine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.