missing translation for 'onlineSavingsMsg'
Learn More

Trimetazidine dihydrochloride, MedChemExpress

Product Code. 16408648
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16408648

Brand: Medchem Express HYB0968

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Trimetazidine dihydrochloride is a selective long chain 3-ketoyl coenzyme A thiolase inhibitor with an IC50 of 75 nM, which can inhibit β-oxidation of free fatty acid (FFA).

Trimetazidine dihydrochloride is an effective antianginal agent and a cytoprotective drug, has anti-oxidant, anti-inflammatory, antinociceptive and gastroprotective properties. Trimetazidine dihydrochloride triggers autophagy. Trimetazidine dihydrochloride is also a 3-hydroxyacyl-CoA dehydrogenase (HADHA) inhibitor.

Specifications

Chemical Name or Material Trimetazidine dihydrochloride
CAS 13171-25-0
Color White
Purity Grade Notes Research
Molecular Formula C14H24Cl2N2O3
Quantity 10 mg
Solubility Information H2O : ≥ 100 mg/mL (294.76 mM) ∣DMSO : 25 mg/mL (73.69 mM; Need ultrasonic)
SMILES COC1=CC=C(CN2CCNCC2)C(OC)=C1OC.[H]Cl.[H]Cl
Molecular Weight (g/mol) 339.26
Formula Weight 339.26
Percent Purity 99.62%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.