Learn More
Triphenylphosphine dibromide, 96%, Thermo Scientific Chemicals
41.21 € - 620.09 €
Chemical Identifiers
CAS | 1034-39-5 |
---|---|
Molecular Formula | C18H15Br2P |
Molecular Weight (g/mol) | 422.10 |
MDL Number | MFCD00000054 |
InChI Key | OCXGTPDKNBIOTF-UHFFFAOYSA-N |
Synonym | dibromotriphenylphosphorane, phosphorane, dibromotriphenyl, triphenyldibromophosphorane, unii-7y0j6r63jd, bromotriphenylphosphonium bromide, dibromotriphenyl-??-phosphane, dibromotriphenylphosphine, dibromtriphenylphosphorane, triphenylphosphinedibromide, dibromo triphenylphosphoraneShow More |
PubChem CID | 70579 |
IUPAC Name | dibromo(triphenyl)-$l^{5}-phosphane |
SMILES | BrP(Br)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11414593
|
Thermo Scientific Alfa Aesar
A11552.09 |
10 g |
41.21 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11424593
|
Thermo Scientific Alfa Aesar
A11552.18 |
50 g |
149.75 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11434593
|
Thermo Scientific Alfa Aesar
A11552.30 |
250 g |
620.09 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Triphenylphosphine dibromide is used as a reagent for conversion of aldoximes into nitriles. It acts as a reagent for cleavage of acetals. It reacts with amines to give iminophosphoranes. Further, it serves as a reagent in the preparation of alkyl and aryl bromides from alcohols and phenols without rearrangement.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTriphenylphosphine dibromide is used as a reagent for conversion of aldoximes into nitriles. It acts as a reagent for cleavage of acetals. It reacts with amines to give iminophosphoranes. Further, it serves as a reagent in the preparation of alkyl and aryl bromides from alcohols and phenols without rearrangement.
Solubility
Soluble in water, dichloromethane, acetonitrile, phenyl cyanide, dimethyl formamide, ether, chloroform, alcohol, carbon tetrachloride, acetic acid and carbon disulfide. Sparingly soluble in phenyl chloride, triphenylphosphine and phenyl hydride. Insoluble in aqueous bromine.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong bases, strong oxidizing agents, bases and active metals.
Chemical Identifiers
1034-39-5 | |
422.10 | |
OCXGTPDKNBIOTF-UHFFFAOYSA-N | |
70579 | |
BrP(Br)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C18H15Br2P | |
MFCD00000054 | |
dibromotriphenylphosphorane, phosphorane, dibromotriphenyl, triphenyldibromophosphorane, unii-7y0j6r63jd, bromotriphenylphosphonium bromide, dibromotriphenyl-??-phosphane, dibromotriphenylphosphine, dibromtriphenylphosphorane, triphenylphosphinedibromide, dibromo triphenylphosphoraneShow More | |
dibromo(triphenyl)-$l^{5}-phosphane |
Specifications
1034-39-5 | |
C18H15Br2P | |
UN3261 | |
Moisture sensitive | |
Soluble in water,dichloromethane,acetonitrile,phenyl cyanide,dimethyl formamide,ether,chloroform,alcohol,carbon tetrachloride,acetic acid and carbon disulfide. Sparingly soluble in phenyl chloride,triphenylphosphine and phenyl hydride. Insoluble in aqueous bromine. | |
BrP(Br)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
422.10 | |
422.1 | |
Triphenylphosphine dibromide |
∼260°C (decomposition) | |
MFCD00000054 | |
917652 | |
dibromotriphenylphosphorane, phosphorane, dibromotriphenyl, triphenyldibromophosphorane, unii-7y0j6r63jd, bromotriphenylphosphonium bromide, dibromotriphenyl-??-phosphane, dibromotriphenylphosphine, dibromtriphenylphosphorane, triphenylphosphinedibromide, dibromo triphenylphosphorane | |
OCXGTPDKNBIOTF-UHFFFAOYSA-N | |
dibromo(triphenyl)-$l^{5}-phosphane | |
70579 | |
96% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P370+P380-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 213-855-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.