missing translation for 'onlineSavingsMsg'
Learn More

Triphenylsilanol, 98%

Product Code. p-7024505
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11413900

Brand: Thermo Scientific Alfa Aesar A19284.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 791-31-1 | C18H16OSi | 276.41 g/mol

Triphenylsilanol is used as a water surrogate for regioselective Pd catalyzed allylations. A thick on germanium substrates have been produced by the electron bombardment of an evaporated thin film of triphenylsilanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylsilanol is used as a water surrogate for regioselective Pd catalyzed allylations. A thick on germanium substrates have been produced by the electron bombardment of an evaporated thin film of triphenylsilanol.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 791-31-1
Molecular Formula C18H16OSi
Molecular Weight (g/mol) 276.41
MDL Number MFCD00002102
InChI Key NLSXASIDNWDYMI-UHFFFAOYSA-N
Synonym triphenylsilanol, silanol, triphenyl, triphenylhydroxysilane, silane, hydroxytriphenyl, unii-azc8o4ttx4, silanol, 1,1,1-triphenyl, hydroxy triphenyl silane, si oh ph3, azc8o4ttx4
PubChem CID 69925
ChEBI CHEBI:35035
IUPAC Name hydroxy(triphenyl)silane
SMILES C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)O

Specifications

Melting Point 149°C to 154°C
Quantity 5 g
Beilstein 985007
Solubility Information Insoluble in water.
Formula Weight 276.41
Percent Purity 98%
Sensitivity Moisture sensitive
Density 1.17
Chemical Name or Material Triphenylsilanol
compliance-icons
Product Identifier
  • Triphenylsilanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : triphenylsilanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.