missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TTLL3 Recombinant Protein

335.00 € - 508.00 €

Specifications

Accession Number AAH09479
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 26140
Molecular Weight (g/mol) 36.85
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16128122
View Documents Promotion Details
Abnova™
H00026140-P01.10ug
10 ug
335.00 €
10µg
Please to purchase this item. Need a web account? Register with us today!
16138122
View Documents Promotion Details
Abnova™
H00026140-P01.25ug
25 ug
508.00 €
25µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH09479
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
36.85
8
Glutathione Sepharose 4 Fast Flow
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TTLL3
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
26140
TTLL3 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
TEGDRNIWIVKPGAKSRGRGIMCMDHLEEMLKLVNGNPVVMKDGKWVVQKYIERPLLIFGTKFDLRQWFLVTDWNPLTVWFYRDSYIRFSTQPFSLKNLDK
DKFZp434B103/DKFZp686D076/FLJ13898/HOTTL/MGC120529/MGC120530/MGC120532
TTLL3
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Product Certifications