Learn More
Invitrogen™ Tubulin Tracker™ Green (Oregon Green™ 488 Taxol, bis-acetate)
Description
Tubulin Tracker Green is an uncharged, non-fluorescent compound that easily passes through the plasma membrane of live cells. Once inside the cell, the lipophilic blocking group is cleaved by non-specific esterases, resulting in a green-fluorescent, charged form, inhibiting it from escaping the live cells. TubulinTracker Green reagent utilizes Oregon Green 488 dye, which absorbs and emits optimally at 494 nm and 522 nm, respectively, and fluorescence can be visualized with standard GFP/FITC filter settings using almost any fluorescent imaging instrument. It can be multiplex with blue, orange, red, deep-red, and near-IR fluorophores.
Specifications
Specifications
| Content And Storage | Contains sufficient material for at least 60 slides at 1 mL / slide and 1 x working concentration. 1 x 20 μg Tubulin Tracker Green (Oregon Green 488 Taxol, bis-acetate) 1 x 1 mL Pluronic F-127, 20% solution in DMSO |
| Excitation Wavelength Range | 494/522 |
| Dye Type | Oregon Green™ 488 |
| Quantity | 60 slides |
| Product Line | TubulinTracker |
| Shipping Condition | Approved for shipment on Wet or Dry Ice |
| Label Type | UV-Vis Labels |
| Product Type | Tubulin Tracker |
| SubCellular Localization | Tubulin |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.