Learn More
Fisherbrand™ Turbo PTFE Stir Bar Retriever with Rare Earth Core
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
350 mm
450 mm
Unit Size:
Each
Spezifikation
Spezifikation
| Color | White |
| Description | Turbo PTFE Stir Bar Retriever with Rare Earth Core for Giant Stir Bars |
| For Use With (Equipment) | For the removal of Giant Stir Bars from vessels of all kinds |
| Diameter (Metric) | 18 mm |
| Length (Metric) | 350 mm |
| Material | PTFE with Rare Earth Magnet Core |
| Quantity | 1 |