missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TYMP/CEN22q FISH Probe

Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.

1515.00 € - 2495.00 €

Specifications

Content And Storage Store at 4°C in the dark.
Description DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Format Tube
For Use With (Application) FISH-Ce
Gene ID (Entrez) 1890
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16093275
View Documents
Abnova™
FG0179.100uL
100 μL
1515.00 €
100µL
Please to purchase this item. Need a web account? Register with us today!
16003285
View Documents
Abnova™
FG0179.200uL
200 μL
2495.00 €
200µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Store at 4°C in the dark.
Tube
1890
Liquid
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
FISH-Ce
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Videos
SDS
Documents

Documents

Product Certifications