Learn More
DuPont™ Tychem™ 2000 C Boot cover
Shop All DuPont Products
Click to view available options
Size:
Universal
Description
- Partially stitched, partially taped
- Slip-retardant sole
- Knee-length with fixation ties
- Yellow
- Certified according to Regulation (EU) 2016/425
- Partial body chemical protective clothing, Category III, Type PB [3]
- EN 14126 (barrier to infective agents)
- Antistatic treatment (EN 1149-5) - on inside
Specifications
Specifications
Size | Universal |
Color | Yellow |
Sole Type | Slip-resistant |
Seam Type | Partiall stitched, partially taped |
Includes | Fixation ties |
Certifications/Compliance | (EU) 2016/425, Category III, Type PB [3], EN 14126, EN 1149-5 |
Product Type | Boot Covers |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DuPont™ Tychem™ 2000 C Boot cover >
Spot an opportunity for improvement?Share a Content Correction