Learn More
Paragon Scientific™ Ubbelohde Viscometer for Tranparent Liquids
For the determination of kinematic viscosity of transparent Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
Brand:  Paragon Scientific™ UBBEL11NC
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 16040490
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
Specifications
600 to 3000 cSt
3
ASTM D 445, D 446 and ISO 3104, 3105
No
D 445, D 446
3C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
4 week lead-time