missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULK3 293T Cell Overexpression Lysate (Denatured), Abnova
Brand: Abnova H00025989-T01.100uL
Additional Details : Weight : 0.00010kg
Description
Western Blotting
Specifications
Western Blot | |
ULK3 293T Cell Transient Overexpression Lysate(Denatured) | |
23.65 | |
pCMV-ULK3 full-length | |
100 μL | |
Human |
293T | |
BC056423 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-ULK3 antibody (H00025989-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |