Learn More
Advantage-Lab™ Ultrasonic Bath Beakers with Covers
Product Code. 15628805
Encompass
Click to view available options
For Use With (Equipment):
AL04-01-230 Ultrasonic Water Bath
AL04-02-230 Ultrasonic Water Bath
AL04-03-230 Ultrasonic Water Bath
AL04-04-230 Ultrasonic Water Bath
AL04-06-230 Ultrasonic Water Bath
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 15628805
Brand: Advantage-Lab™ AL0404152
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
Isolate items for ultrasonification in these covered beakers. Advantage-Lab™ Ultrasonic Bath Beakers with Covers make it easier to treat small and individual items.
  • Covers provided.
  • Cover with holes provided.
  • Rubber ring provided.
  • Set of 2.
Specifications
For Use With (Equipment) AL04-04-230 Ultrasonic Water Bath
Weight (Metric) 2.0 kg
Includes Covers, cover with holes, and rubber ring.
Quantity 2/Pk.