missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC 0631, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Description
UNC 0631 is a potent histone methyltransferase G9a inhibitor with an IC50 of 4 nM. UNC 0631 potently reduces H3K9me2 levels in MDA-MB-231 cells with an IC50 of 25 nM.
Specifications
Specifications
| Chemical Name or Material | UNC 0631 |
| CAS | 1320288-19-4 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C37H61N7O2 |
| Quantity | 5 mg |
| Solubility Information | DMF : 150 mg/mL (235.88 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (157.25 mM; Need ultrasonic) ∣DMSO : 16.67 mg/mL (26.21 mM; Need ultrasonic) |
| SMILES | COC1=CC2=C(NC3CCN(CC4CCCCC4)CC3)N=C(N5CCN(C(C)C)CCC5)N=C2C=C1OCCCN6CCCCC6 |
| Molecular Weight (g/mol) | 635.93 |
| Formula Weight | 635.93 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction