Learn More
UNC 0631, MedChemExpress
MedChemExpress UNC 0631 is a potent histone methyltransferase G9a inhibitor with an IC50 of 4 nM. UNC 0631 potently reduces H3K9me2 levels in MDA-MB-231 cells with an IC50 of 25 nM.
Brand: Medchem Express HY-13808
Description
UNC 0631 is a potent histone methyltransferase G9a inhibitor with an IC50 of 4 nM. UNC 0631 potently reduces H3K9me2 levels in MDA-MB-231 cells with an IC50 of 25 nM.Specifications
UNC 0631 | |
Yellow | |
C37H61N7O2 | |
DMF : 150 mg/mL (235.88 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (157.25 mM; Need ultrasonic) ∣DMSO : 16.67 mg/mL (26.21 mM; Need ultrasonic) | |
635.93 | |
99.35% | |
Solid |
1320288-19-4 | |
Research | |
25 mg | |
COC1=CC2=C(NC3CCN(CC4CCCCC4)CC3)N=C(N5CCN(C(C)C)CCC5)N=C2C=C1OCCCN6CCCCC6 | |
635.93 | |
Research | |
Cancer-programmed cell death |