Learn More
UNC 0631, MedChemExpress
MedChemExpress UNC 0631 is a potent histone methyltransferase G9a inhibitor with an IC50 of 4 nM. UNC 0631 potently reduces H3K9me2 levels in MDA-MB-231 cells with an IC50 of 25 nM.
Brand:  Medchem Express HY-13808
Encompass_Preferred
 View more versions of this product
Product Code. 16473386
  • 530.55 € / 25mg
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
UNC 0631 is a potent histone methyltransferase G9a inhibitor with an IC50 of 4 nM. UNC 0631 potently reduces H3K9me2 levels in MDA-MB-231 cells with an IC50 of 25 nM.
Specifications
Specifications
UNC 0631
Yellow
C37H61N7O2
DMF : 150 mg/mL (235.88 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (157.25 mM; Need ultrasonic) ∣DMSO : 16.67 mg/mL (26.21 mM; Need ultrasonic)
635.93
99.35%
Solid
1320288-19-4
Research
25 mg
COC1=CC2=C(NC3CCN(CC4CCCCC4)CC3)N=C(N5CCN(C(C)C)CCC5)N=C2C=C1OCCCN6CCCCC6
635.93
Research
Cancer-programmed cell death
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
UNC 0631, MedChemExpress