Learn More
Tocris Bioscience™ UNC 2327
Allosteric inhibitor of PRMT3
Brand: Tocris Bioscience™ 5057/10
228.07 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
UNC 2327 is an allosteric inhibitor of protein arginine methyltransferase 3 (PRMT3) (IC50 = 230 nM). Noncompetitive with both the peptide substrate and cofactor.Specifications
>99% | |
319.38 | |
10 mg |
UNC 2327 | |
UNC 2327 |