Learn More
Thermo Scientific™ UNC1999, 95%
Brand: Thermo Scientific™ 468221000
563.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
UNC1999 | |
100 mg | |
569.75 | |
Reagent |
1431612-23-5 | |
C33 H43 N7 O2 | |
Solid | |
95% |