Learn More
Haldenwanger™ Unglazed Porcelain Pestle
Use these pestles for a variety of laboratory applications. Haldenwanger™ Unglazed Porcelain Pestles are constructed from optimized laboratory porcelain.
Brand: Haldenwanger™ L 56/3/RAUH
This item is not returnable.
View return policy
Description
- Available in a variety of sizes
- Grinding surfaces are unglazed
- Complies with DIN VDE 0335, type C 100
Specifications
Unglazed | |
Pestle | |
Porcelain | |
175mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haldenwanger™ Unglazed Porcelain Pestle > Length: 175mm
Spot an opportunity for improvement?Share a Content Correction