Learn More
Marienfeld Superior™ UniMark™ Uncoated Microscope Slides with Printed Marking Area
Marienfeld Superior™ UniMark™ miscroscope slides have a printed area which can take notes printed by most types of printers and can be written with permanent markers. Different colors of the labelling area offer the possibility to distinct the preparations (by users, etc.)
Brand:  Marienfeld Superior™ 0704002
 View more versions of this product
Product Code. 10285232
  • 10.00 € / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made of soda lime glass of third hydrolytic class
  • In compliance with DIN ISO 8037/1
  • Pre-cleaned, ready for use, autoclavable
  • With printed marking area in different colors
  • 50 pieces in a box, 50 boxes in a carton
  • For in-vitro diagnostic (IVD) applications according to IVD directive 98/79/EC, with CE-mark, recommended best-before-date and batch number for comprehensive information and traceability
Specifications
Specifications
76 mm
2500
90°, Ground
Soda Lime Glass Type III, Hydrolytic Class
1 mm
White
UniMark™ Microscope Slides with printed marking area
76 x 26 mm
26 mm
50
Yes
Yes
Microscope Slide
CE Mark, DIN ISO 8037/1
50 Pieces in Cardboard Box