Learn More
Grant Instruments™ Universal Tray for Grant Shaking Water Baths
Product Code. 15355812
Click to view available options
For Use With (Equipment):
LSB12
LSB18
OLS26
Unit Size:
Each
Product Code. 15355812
Brand: Grant Instruments™ TU26
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
Holds flasks, bottles, and other vessels in place when used with Grant Shaking Water Baths. Includes adjustable springs.
  • Universal tray with adjustable springs
  • Highly versatile for a variety of vessel types
TRUSTED_SUSTAINABILITY
Specifications
Description TU26 universal tray with adjustable springs to accommodate a variety of vessel types for OLS26
For Use With (Equipment) OLS26
Type Universal Tray
Weight (Metric) 2500 g