Learn More
Kinesis™ Low-Pressure Unions: Kel-F
Click to view available options
Material:
Kel-F
Unit Size:
Each
Description
- Manufactured from inert polymers
- Provide your own fittings to suit your application
- 1/4-28 flat-bottom male thread union
- 0.062 in. thru-hole diameter
- PCTFE body
Specifications
Specifications
| Type | Union |
| Temperature (Metric) Max | 80°C |
| Material | Kel-F |
| Color | Natural |