missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uridine 5'-monophosphate, disodium salt, hydrate, 98%, Thermo Scientific Chemicals
homogeneous
53.34 € - 342.71 €
Chemical Identifiers
CAS | 3387-36-8 |
---|---|
Molecular Formula | C9H11N2Na2O9P |
Molecular Weight (g/mol) | 368.15 |
MDL Number | MFCD00149422 |
InChI Key | KURVIXMFFSNONZ-IJRYJGAHNA-L |
Synonym | uridine-5'-monophosphate disodium salt, uridine 5'-monophosphate, disodium salt, 5'-uridylicaciddisodiumsalt |
PubChem CID | 123134060 |
SMILES | [Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])([O-])=O)N1C=CC(=O)NC1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10052890
|
Thermo Scientific Acros
226290010 |
1 g | Glass bottle |
53.34 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10762651
|
Thermo Scientific Acros
226290050 |
5 g | Glass bottle |
90.95 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15760839
|
Thermo Scientific Acros
226290250 |
25 g | Glass bottle |
342.71 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3387-36-8 | |
368.15 | |
KURVIXMFFSNONZ-IJRYJGAHNA-L | |
123134060 |
Specifications
3387-36-8 | |
208.0°C to 210.0°C | |
Authentic | |
C9H11N2Na2O9P | |
10, 9689 | |
Solubility in water:+/- 40% (20°C) | |
KURVIXMFFSNONZ-IJRYJGAHNA-L | |
disodium [(2R,3S,4R,5R)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate | |
368.15 | |
368.14 | |
Crystalline Powder |
0.71 to 0.77 (A250/A260),0.38 to 0.44 (A280/A260) | |
White | |
97.5% min. (HPLC) | |
MFCD00149422 | |
uridine-5'-monophosphate disodium salt, uridine 5'-monophosphate, disodium salt, 5'-uridylicaciddisodiumsalt | |
97.5% min. (lambda max. 262nm,pH 7) molar absorptivity 10000+/-2%,on dry basis | |
[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])([O-])=O)N1C=CC(=O)NC1=O | |
95% min. (c=1, 430nm) 1cm cell | |
123134060 | |
98% | |
Uridine 5′-monophosphate, disodium salt, hydrate |
Safety and Handling
missing translation for 'einecsNumber' : 222-211-9
Product Identifier
- Uridine 5'-monophosphate, disodium salt, hydrate
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Uridine 5'-monophosphate, disodium salt, hydrate, 98%, Thermo Scientific Chemicals