missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ USB-Ethernet Adapter for UN260plus

USB-Ethernet Adapter accessory for Memmert Ovens.

Brand:  Memmert™ UN260plusE06192

Product Code. 17570016

  • 56.40 € / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Accessory
  • For use with Memmert Ovens
Specifications

Specifications

UN260plus Oven
USB Ethernet Adapter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Memmert™ USB-Ethernet Adapter for UN260plus > For Use With: UN260plus

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.