Learn More
Advantage-Lab™ UV Lamp, 1 x 8 Watt
Useful for detecting fluorescent particles and minerals, curing adhesives and paints, and testing coatings. Advantage-Lab™ UV Lamp 1 x 8 Watt is a high-intensity, single-wave unit designed with an integrated handle and a powder-coated, metal housing.
Marque:  Advantage-Lab™ AL10-08-254
Encompass
 Afficher plus de versions de ce produit
Code produit. 15648885
  • 586.00 € / Each
Estimated Shipment: 07-01-2025
to see stock.
Découvrez plus d'offres spéciales
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
  • Useful in many fields including the following:
    • Molecular biology.
    • Microbiology.
    • Botany.
    • Chemistry.
    • Mineralogy.
    • Glass production.
  • High intensity.
  • Single-wave unit.
  • Powder-coated metal housing.
  • Integrated handle.
  • Adjustable stand (option).
Spécification
Spécification
220/240 V (optional 110/120 V)
Shortwave UV-C
680 μW/cm2
1 x 8 W
335 x 70 x 65 mm
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Certificats