Learn More
Uvex™ Disposable Chemical Protection Arm Warmers
Click to view available options
Includes: 50
Description
Specifications
Specifications
Size | Universal |
Product Type | General Purpose Sleeve |
Material | Non-woven Spunbond Polypropylene, Polyethylene Laminate Film |
Disposable | Single-use |
Weight (Metric) | 60 g/m² |
Packaging | 50/Pk. |