Learn More
UVP UVP Ultra Violet Product UV Viewing Cabinet
Brand:  UVP 95-0253-02
Product Code. 11718211
  • 3385.00 € / Each
Only null left
Estimated Shipment: 01-09-2025
Add to basket
Specifications
230V
400mm
365nm
C-75
1Pack
419mm
2X
602mm
254-365nm
UV Viewing Cabinet
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
UVP UVP Ultra Violet Product UV Viewing Cabinet > Depth: 419mm