Learn More
BD Vacutainer™ Safety-Lok™ Blood Collection Set
Designed to help prevent needle stick injuries. BD™ Vacutainer™ Safety-Lok™ Blood Collection Set are sterile, closed systems for venous blood collection. Each versatile winged infusion set includes an irreversible safety mechanism that can be activated with one hand.
Brand:  BD 367295
 View more versions of this product
Product Code. 15940821
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
Safety-Lok™ blood collection sets are sterile, closed systems for venous blood collection. They can also be used for short-term infusions of up to two hours. These winged infusion sets include a hypodermic needle, color-coded wings, an integrated safety shield, small-bore tubing and a connector. Many also include a Luer adapter.
  • Safety mechanism protects against needle injuries following successful venipuncture
  • Safety shield pushes over the needle, surrounding it completely
  • Safety mechanism engages irreversibly with an audible click over the needle
  • Single-handed activation so attention remains focused on the patient and the puncture site
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Blood Collection Set
25 G
Venous blood collection, Short-term infusions
Yes
178 mm
Dark Blue
Sterile
With luer adapter, integrated safety shield
19 mm
200 Cs.