Learn More
Valrocemide, MedChemExpress
MedChemExpress Valrocemide (TV1901) is a promising antiepileptic drug candidate that shows a broad spectrum of anticonvulsant activity.

POA - 132.30 €
Specifications
Chemical Name or Material | Valrocemide |
---|---|
Physical Form | Solid |
CAS | 92262-58-3 |
Color | White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16455717
![]() |
Medchem Express
HY-100379 |
5 mg |
POA
5mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16442566
![]() |
Medchem Express
HY-100379 |
25 mg |
58.05 €
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16439546
![]() |
Medchem Express
HY-100379 |
50 mg |
87.75 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16432978
![]() |
Medchem Express
HY-100379 |
10 mg |
35.10 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16414296
![]() |
Medchem Express
HY-100379 |
100 mg |
132.30 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Valrocemide (TV1901) is a promising antiepileptic drug candidate that shows a broad spectrum of anticonvulsant activity.Specifications
Valrocemide | |
92262-58-3 | |
Research | |
Neuroscience-Neuromodulation | |
DMSO : ≥ 100 mg/mL (499.30 mM) | |
200.28 | |
98.42% |
Solid | |
White | |
C10H20N2O2 | |
TV1901 | |
CCCC(CCC)C(NCC(N)=O)=O | |
200.28 | |
Research |
Safety and Handling
H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.