Learn More
B Braun™ Vasofix™ Safety Shielded IV Catheter with Injection Port
Protects you and your healthcare team against needlestick injuries and related infections.
Brand:  B Braun™ 4269071S-01
Product Code. 10323419
  • 90.00 € / Each
Please to purchase this item. Need a web account? Register with us today!
Edge
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • IV catheter material of polyurethane
  • Integrated port allowing injections without an extra needle or risk of needlestick injuries
  • Sharp backcut bevel for minimal puncture trauma
  • Transparent catheter segments provide confirmation of successful catheter placement when withdrawing the needle
  • Latex- and PVC-free
  • According to ISO 10.555-1/5
  • Color coded yellow
  • Size of 24 G, diameter of 0.7 x 19 mm
  • Box of 50 pieces
Specifications
Specifications
IV Catheter
24 G
Needlestick injury protection
50/Pk.
Yellow
ISO-Standard 10.555-1/5
19 mm