Learn More
Thermo Scientific™ Versette™ ControlMate™ Software
Simplify creation and execution of pipetting programs.
Brand: Thermo Scientific™ 650-09-CMSW
Additional Details : Weight : 0.20000kg
Description
Simplify creation and execution of pipetting programs with the Thermo Scientific™ Versette™ ControlMate™ Software, an easy and straightforward way to create and run pipetting programs with the Versette automated liquid handler.
Programs quickly for easy liquid transfer operations or for more complex pipetting sequences like serial dilutions, plate reformatting or immunoaffinity sample preparations with MSIA pipette tips. This PC software includes a wide labware library and precise controlling options for liquid handling applications.
Versette ControlMate Software 1.2.0
- Easy to use and easy to get started
- Simple operation with basic commands or complex protocols with extended command list
- Wide labware library easily modified by the user
- Complete control using simple commands in a tree structure
Specifications
Automated Liquid Transfer Software | |
Versette ControlMate Software | |
Windows XP™, Windows Vista™ (32-and 64-bit), Windows 7™ (32- and 64-bit) |