Learn More
Thermo Scientific™ Viper™ and nanoViper™ Fingertight Fittings Accessories
Obtain reproducible chromatographic results using tool-free and easy-to-use Thermo Scientific™ Viper™ and Thermo Scientific™ nanoViper™ Fingertight Fittings.
317.00 €
Specifications
Type Accessory
View More Specs
Products 1
Product Code Brand Description Material Product Line For Use With (Application) Price Quantity & Availability  
Product Code Brand Description Material Product Line For Use With (Application) Price Quantity & Availability  
15926465
View Documents
Thermo Scientific™
6036.1045
Viper Inline Filter Titanium Viper Column Protection
317.00 €
Each
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
Description
Description
Ensure optimal separations and maximum LC performance using virtually zero-dead-volume connections with the Thermo Scientific™ Viper™ Fingertight Fitting systems. Accessories help complete the installation according to your application needs.
See Viper and nanoViper resources
We offer the following accessories for Viper Fingertight Fittings Systems:
  • Union exclusively designed for connecting Viper and nanoViper Fingertight Fittings directly without additional dead volume
  • Viper Plug for installation in open valve positions to protect the bores or for diagnostic functions
  • Tool to recalibrate Viper and nanoViper fittings for an extended lifetime when connected to multiple columns with non-standard column bore geometry
  • Viper Inline Filter (titanium, av. frit pre size 0.5μm) to protect the column from contaminants
  • Adaptor for Viper Fingertight Fittings (2 pcs, stainless steel, for columns ≥ 2.1 mm ID) to adapt to non-standard column bore geometry
Warranty
Viper and nanoViper Fingertight Fittings Accessories
Specifications
Specifications
Accessory
Videos
SDS
Documents
Documents
Product Certifications