Learn More
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Amber Graduations
Product Code. 10456691
Click to view available options
Volume (Metric):
1 mL
3 mL
4 mL
5 mL
10 mL
15 mL
20 mL
50 mL
100 mL
Unit Size:
Each
Pack of 5
This item is not returnable. View return policy
Product Code. 10456691
Brand: Poulten & Graf GmbH 1 2323004
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Amber Graduations allow accurate measurements of a volume of solution.
  • With 2 marks
  • Amber graduations
  • Conformity approved
  • With batch certificate
Specifications
Certifications/Compliance EN ISO 648
Class AS
Material Soda Lime Glass
Sterility Non-sterile
Graduated Yes
Graduation Color Amber
Graduation Type Ring marks
Quantity 5
Tolerance 0.015 mL
Volume (Metric) 4 mL
Type Two-mark Bulb Pipette
Disposable Reusable