Learn More
Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Blue Graduation
Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Blue Graduation is ideal for measuring and transferring various quantities of liquid.
Brand: Poulten & Graf GmbH 1 11437402
This item is not returnable.
View return policy
Description
Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Blue Graduation is ideal for measuring and transferring various quantities of liquid.
- Total delivery
- Blue graduation
- With ring marks
- Comformity approved
Specifications
EN ISO 835 | |
Graduated pipette | |
Non-sterile | |
0.05 mL | |
Graduated Pipet |
AS | |
Soda Lime Glass | |
Yes | |
10 mL | |
Reusable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Blue Graduation > Capacity, Metric: 10mL
Spot an opportunity for improvement?Share a Content Correction