Learn More
VX-984, MedChemExpress
MedChemExpress VX-984 (M9831) is an orally active, potent, selective and BBB-penetrated DNA-PK inhibitor. VX-984 efficiently inhibits NHEJ (non-homologous end joining) and increases DSBs (DNA double-strand breaks). VX-984 can be used for glioblastomas (GBM) and non-small cell lung cancer (NSCLC) research.
Brand: Medchem Express HY-19939S
Description
VX-984 (M9831) is an orally active, potent, selective and BBB-penetrated DNA-PK inhibitor. VX-984 efficiently inhibits NHEJ (non-homologous end joining) and increases DSBs (DNA double-strand breaks). VX-984 can be used for glioblastomas (GBM) and non-small cell lung cancer (NSCLC) research.Specifications
VX-984 | |
50 mg | |
C23H21D2N7O | |
Solid | |
White | |
O=C(C1=CC=NC2=C([C@H](C)CNC3=CC(C4=C([2H])N=C(C)N=C4[2H])=NC=N3)C=CC=C12)NC | |
415.49 | |
Research |
1476074-39-1 | |
Research | |
Cancer-Kinase/protease | |
M9831 | |
DMSO : 10 mg/mL (24.07 mM; Need ultrasonic) | |
415.49 | |
98.34% |