Learn More
Saint Gobain Life Sciences Chemware™ PFA Graduated Wide Mouth Wash Bottle
Click to view available options
Capacity (Metric):
100 mL
250 mL
1000 mL
Description
- Wide-mouth for easy filling
- Chemically inert and leak-proof
- Flexible nozzle with 360° swivel; small orifice allows accurate content dispensing
- Transparent and break resistant
- Molded-in graduation marks
Specifications
Specifications
Capacity (Metric) | 1000 mL |
Color | Transparent |
Material | PFA |
Neck ID (Metric) | 36.9 mm |
Graduated | Yes |
Mouth | Wide |
Height (Metric) | 317.5 mm |
Product Type | Wash Bottle |
Height (English) | 12.5 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Saint Gobain Life Sciences Chemware™ PFA Graduated Wide Mouth Wash Bottle >
Spot an opportunity for improvement?Share a Content Correction