Learn More
Karl Hecht™ Washing Bottle (Spray)
Product Code. 10003791
Click to view available options
Capacity:
1000mL
This item is not returnable. View return policy
This item is not returnable. View return policy
Constructed of durable Duran™ borosilicate glass 3.3. Karl Hecht™ Washing Bottle (Spray) comes with an interchangeable, NS 29/32 spray tube.
  • Duran™ borosilicate glass 3.3 construction.
  • Interchangeable NS 29/32 spray tube.
Specifications
Capacity 1000mL
Product Type Wash Bottle
Includes Interchangeable, NS 29/32 spray tube
Material Borosilicate Glass (3.3)
Standard Taper Joint NS 29/32