Learn More
Karl Hecht™ Watch Glasses
These watch glasses are useful for evaporating liquids, holding solids, and heating small amounts of a sample.
4.00 €
Specifications
Description Fused rim
Material Glass
Shape Round
View More Specs
Products 1
Product Code Brand Diameter (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Price Quantity & Availability  
10339441
View Documents
Karl Hecht™
40010014
14 cm
4.00 €
Each
Estimated Shipment: 30-12-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12 341
  • With fused rim
  • Manufactured from glass
  • Available in different diameters
Specifications
Specifications
Fused rim
Glass
Round
Videos
SDS
Documents
Documents
Product Certifications