Learn More
Cytiva Whatman™ Cellulose Nitrate Membrane Filters
Manufactured under strictly controlled conditions. Whatman™ Cellulose Nitrate Membrane Filters have a noticeably improved flexibility and are made to tolerate abuse during handling, loading, and autoclaving without sacrificing integrity.
108.24 € - 263.00 €
Specifications
Format | Disc |
---|---|
Material | Cellulose Nitrate |
Quantity | 100 Pack |
Thickness (Metric) | 105 to 140 μm |
Product Code | Brand | Diameter (Metric) | Pore Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Pore Size | Price | Quantity & Availability | ||||
11343135
|
Cytiva
10400014 |
50 mm | 12 μm |
246.00 €
Pack of 100 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
11363135
|
Cytiva
10400106 |
25 mm | 8 μm |
174.00 €
Pack of 100 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
11383135
|
Cytiva
10400112 |
47 mm | 8 μm |
238.00 €
Pack of 100 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
11345284
|
Cytiva
10400206 |
25 mm | 5 μm |
174.00 €
Pack of 100 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
10353101
|
Cytiva
10400212 |
47 mm | 5 μm |
238.00 €
Pack of 100 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
11365284
|
Cytiva
10400214 |
50 mm | 5 μm |
246.00 €
Pack of 100 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
11325164
|
Cytiva
10401306 |
25 mm | 0.2 μm | ||||||
11395154
|
Cytiva
10401312 |
47 mm | 0.2 μm |
219.00 €
Pack of 100 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
11305164
|
Cytiva
10401314 |
50 mm | 0.2 μm |
228.00 €
Pack of 100 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
11385154
|
Cytiva
10402014 |
50 mm | 0.1 μm |
234.00 €
Pack of 100 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
11375154
|
Cytiva
10402012 |
47 mm | 0.1 μm |
227.00 €
Pack of 100 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
11304864
|
Cytiva
7191-005 |
50 mm | 1.2 μm |
222.00 €
Pack of 100 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
10719581
|
Cytiva
10400012 |
47 mm | 12 μm |
263.00 €
Pack of 100 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
11393135
|
Cytiva
10400114 |
50 mm | 8 μm |
246.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Among the strongest of their type available, as measured and compared by burst pressure tests.
- Narrow pore size distribution for improved surface capture and analysis.
- Low levels of extractables to ensure sample integrity.
- High strength and flexibility
- Increased temperature stability
- Reduced shrinkage
Specifications
Disc | |
Cellulose Nitrate | |
100 Pack | |
105 to 140 μm |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- AE100 Membrane, Cellulose nitrate, 12 µm 50 mm, 100 pack
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.