Learn More
GE Healthcare Whatman™ Klari-Flex™ Bottle-top Filtration System
Provide vacuum filtration of aqueous solutions such as cell culture media, biological fluids, and buffers
Brand:  GE Healthcare 65145002
Product Code. 11958149
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Drop-Connect technology ensures fail-safe vacuum connection and unit placement in base.
  • Weighted pedestal base and wide, low-profile funnel/bottle filtration units are stable and tip resistant, minimizing the risk of spills and contamination.
  • Graduation marks face front for readability.
  • Large bottle cap accommodates labeling information.
  • Ergonomically designed bottle and bottle cap provide improved handling.
  • Membrane material is PES.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Funnel and bottle
500 mL