Learn More
Cytiva Whatman™ PTFE Membrane Circles
Product Code. p-4520453
Click to view available options
Pore Size:
0.2 μm
0.45 μm
1.0 μm
5 μm
Diameter (Metric):
25 mm
37 mm
47 mm
50 mm
90 mm
150 mm
Unit Size:
Pack of 25
Pack of 50
Product Code. 11383435
Brand: Cytiva 10411311
Provides clarification of corrosives, solvents, and aggressive fluids. Whatman™ PTFE Membrane Circles are particularly useful in HPLC analysis when solid particles can cause permanent damage to the column.
  • Hydrophobic
  • Chemically stable and inert
  • Can be used up to 120°C
  • Laminated onto randomly arranged polypropylene support material
  • For filtration with organic and chemically aggressive solutions or gases
  • Particularly suitable for HPLC analysis
Specifications
Material PTFE
Pore Size 0.45 μm
Hydrophobicity Hydrophobic
Membrane Type TE 36
No. per Pack 50Pack
Diameter (Metric) 47 mm