Learn More
DWK Life Sciences Wheaton™ Black Phenolic Screw Caps with PTFE Faced 14B White Rubber Liner
Product Code. 12416171 Shop All Wheaton Science Products Products
Click to view available options
Closure Size:
24-400
8-425
Unit Size:
Pack of 200
Product Code. 12416171
Brand: DWK Life Sciences W242711
Please to purchase this item. Need a web account? Register with us today!
Provides completely inert inner seal and surface for sample or product
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material Phenolic
Liner Material TFE, Styrene Butadiene Rubber
Product Type Screw Cap
Closure Size 24-400
Autoclavable Autoclavable