Learn More
DWK Life Sciences Wheaton™ Connection With Black Phenolic Caps
Shop All Wheaton Science Products Products
missing translation for 'orderingAttributeHoverText'
Thread Size:
13-425, 20-400
13-425, 22-400
13-425, 24-400
20-400, 24-400
24-400, 24-400
missing translation for 'unitSize'
Pack of 6
Description
- Inert contact surface
- Easy to assemble
- Connection can be used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
- Phenolic connectors have a working temperature of up to 180°C
- Autoclavable
Spécification
Spécification
| For Use With (Application) | Used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere |
| Product Type | Connector |
| Thread Size | 13-425, 22-400 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu