Learn More
DWK Life Sciences Wheaton™ PTFE Faced Fritted Glass Support Base
For 47mm vacuum filtration assembly
Brand:  DWK Life Sciences 419372
193.10 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 12421581
  • 204.00 € / Each
Estimated Shipment: 31-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
47mm Vacuum Filtration Assembly
Glass, PTFE
Support Base
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences Wheaton™ PTFE Faced Fritted Glass Support Base > Glass support base for Filter Size: 47mm