Learn More
DWK Life Sciences Wheaton™ Solid Top Polypropylene Screw Caps
Shop All Wheaton Science Products Products
Click to view available options
Thread Size (Metric):
13-425
Unit Size:
Pack of 144
Description
- Used with glass bottles or sample vials
- Non-autoclavable
- White colored
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| Quantity | 144/Cs. |
| Thread Size (Metric) | 13-425 |
| Liner Material | Polyvinyl |
| Neck Finish | 39 mm |
| Autoclavable | Not Autoclavable |