Learn More
BRAND™ Wide Neck Glass Erlenmeyer Flasks
Shop All Brand Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
300 mL
500 mL
1000 mL
Unit Size:
Pack of 10
Description
- Manufactured from Boro 3.3 glass
- Wide neck for ease of filling
- Acc. to DIN EN ISO 24450
- Provided in packs of 10
Specifications
Specifications
| Diameter (Metric) Outer Neck | 34 mm |
| Capacity (Metric) | 100 mL |
| Material | Borosilicate Glass (3.3) |
| Height (Metric) | 105 mm |
| No. per Pack | 10 |
| Diameter (Metric) Outer | 64 mm |