Learn More
Kinesis X100 Filtration column TELOS (2 x 20µm PE frits)
Brand:  Kinesis 06479-43
X100 Filtration column TELOS (2 x 20µm PE frits)
Product Code. 12936089
  • ONLINE EXCLUSIVE PRICE
    278.00 €
    222.27 € / Pack of 100
    Save 55.73 €
    20% Off
Only null left
Estimated Shipment: 26-01-2026