Learn More
Epredia X1000 Round Coverslip dia. 13mm #1.5 (0.16-0.19mm)
Brand:  Epredia CB00130RAC20MNZ0
49.19 EUR valid until 2025-12-31
BEST PRICE on Promo! Use promo code "24145" to get your promotional price.
X1000 Round Coverslip dia. 13mm #1.5 (0.16-0.19mm)
Product Code. 11588492
  • ONLINE EXCLUSIVE PRICE
    94.40 €
    84.40 € / Pack of 1000
    Save 10.00 €
    10% Off
View all promotions