missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IGF-I/IGF-1 Quantikine ELISA Kit

Product Code. 18723397
Click to view available options
Quantity:
1 Kit (6 Plates)
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 18723397

Brand: R&D Systems™ SG100B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

From the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

  • A 4.5 hour solid-phase ELISA designed to measure human IGF-1 in cell culture supernates, serum, and plasma.
  • It contains E. coli-expressed recombinant human IGF-1 and has antibodies raised againt the recombinant protein.
  • Results obtained for naturally occurring human IGF-1 showed linear curves that were parallel to the standard curves obtained using the Quantikine kit standards.
  • These results indicate that this kit can be used to determine relative mass values for natural human IGF-1.
  • Intra-Assay Precision (Precision within an assay) Three samples of known concentration were tested twenty times on one plate to assess intra-assay precision
  • Inter-Assay Precision (Precision between assays) Three samples of known concentration were tested in twenty separate assays to assess inter-assay precision. Assays were performed by at least three technicians using two lots of components
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.06 - 4 ng/mL
Assay Sensitivity 0.022 ng/mL
Product Type ELISA
Sample Type Plasma (EDTA), Heparin Plasma, Serum, Serum-free Cell Culture Supernatant
Specificity Natural and Recombinant Human IGF-1
Gene ID (Entrez) 3479
Host Species Human
Quantity 1 Kit (6 Plates)
Storage Requirements Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Synonym IBP1, IGF1, IGF-1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, Somatomedin A, Somatomedin C, Somatomedin-C
Target IGF-I/IGF-1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.