Learn More
Waters Corporation XTerra MS C8 Column
XTerra MS C8 columns are general purpose, hybrid-based, reversed-phase C8 columns that are designed to be compatible with mass spectrometry applications.
Brand:  Waters Corporation 186000483
Encompass
 View more versions of this product
Product Code. 10017951
  • 1207.50 € / Each
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • These first generation columns provide superior pH stability compared to a silica-based column
Specifications
Specifications
4.6 mm
6000 psi (415 Bar)
5 μm
1/Ea.
Yes
50 mm
Spherical
125 Å
L7
Column
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Waters Corporation XTerra MS C8 Column